missing translation for 'onlineSavingsMsg'
Learn More

Gadolinium(III) perchlorate hexahydrate, 50% w/w aq. soln., Reagent Grade, Thermo Scientific Chemicals

Product Code. 11470291
Click to view available options
Quantity:
2 g
10 g
50 g
This item is not returnable. View return policy
This item is not returnable. View return policy

[Gd(DO3A-Pr-ATP)(H2O)2] was synthesized by the reaction of DO3A-Pr-ATP with gadolinium(iii) perchlorate hydrate in water. Gadolinium(III) chelates approved by FDA for human imaging.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14017-52-8
MDL Number MFCD00150239
Synonym gadolinium perchlorate, gadolinium iii perchlorate, acmc-20aku7, gadolinium iii perchlorate solution, gadolinium 3+ triperchlorate, gadolinium perchlorate, crystalline hydrate,, gadolinium iii perchlorate solution, 40 wt. % in h2o

Specifications

Physical Form Liquid
Quantity 2 g
UN Number UN3211
Solubility Information Insoluble in water.
Formula Weight 563.69 (455.60 Anhydrous)
Grade Reagent
Concentration or Composition (by Analyte or Components) 50% w/w aq. soln.
Odor Odorless
Chemical Name or Material Gadolinium(III) perchlorate hexahydrate

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'