missing translation for 'onlineSavingsMsg'
Learn More

Gadolinium(III) sulfate octahydrate, REacton™, 99.9% (REO), Thermo Scientific Chemicals

Cryogenic research

88.04 € - 277.38 €

Chemical Identifiers

CAS 13450-87-8
Molecular Formula Gd2H16O20S3
Molecular Weight (g/mol) 746.788
MDL Number MFCD00149734
InChI Key JBFCVGZGWFIPNK-UHFFFAOYSA-H
Synonym gadolinium iii sulfate octahydrate, digadolinium 3+ octahydrate trisulfate, gadolinium iii sulfate, gadolinium sulfate octahydrate, gadolinium sulfuric acid octahydrate, gadolinium sulfate, gadolinium sulfate-water 2/3/8, gadolinium iii sulfate octahydrate, reacton, gadolinium iii sulfate octahydrate trace metals basis
PubChem CID 16211473
IUPAC Name gadolinium(3+);trisulfate;octahydrate
SMILES O.O.O.O.O.O.O.O.[O-]S(=O)(=O)[O-].[O-]S(=O)(=O)[O-].[O-]S(=O)(=O)[O-].[Gd+3].[Gd+3]
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11360108
View Documents
Thermo Scientific Alfa Aesar
042869.09
10 g
88.04 €
10g
Please to purchase this item. Need a web account? Register with us today!
11350108
View Documents
Thermo Scientific Alfa Aesar
042869.18
50 g
277.38 €
50g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Gadolinium(III) sulfate is used as analytical reagents, in inorganic synthesis and electronic ceramics. It is also useful in cryogenic research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

13450-87-8
746.788
JBFCVGZGWFIPNK-UHFFFAOYSA-H
16211473
O.O.O.O.O.O.O.O.[O-]S(=O)(=O)[O-].[O-]S(=O)(=O)[O-].[O-]S(=O)(=O)[O-].[Gd+3].[Gd+3]
Gd2H16O20S3
MFCD00149734
gadolinium iii sulfate octahydrate, digadolinium 3+ octahydrate trisulfate, gadolinium iii sulfate, gadolinium sulfate octahydrate, gadolinium sulfuric acid octahydrate, gadolinium sulfate, gadolinium sulfate-water 2/3/8, gadolinium iii sulfate octahydrate, reacton, gadolinium iii sulfate octahydrate trace metals basis
gadolinium(3+);trisulfate;octahydrate

Specifications

13450-87-8
Gd2H16O20S3
14,4326
Soluble in cold water
O.O.O.O.O.O.O.O.[O-]S(=O)(=O)[O-].[O-]S(=O)(=O)[O-].[O-]S(=O)(=O)[O-].[Gd+3].[Gd+3]
746.788
746.81 (602.68 Anhydrous)
Reacton™
4.139 g/mL
Gadolinium (III) sulfate octahydrate
Crystalline Aggregates
MFCD00149734
gadolinium iii sulfate octahydrate, digadolinium 3+ octahydrate trisulfate, gadolinium iii sulfate, gadolinium sulfate octahydrate, gadolinium sulfuric acid octahydrate, gadolinium sulfate, gadolinium sulfate-water 2/3/8, gadolinium iii sulfate octahydrate, reacton, gadolinium iii sulfate octahydrate trace metals basis
JBFCVGZGWFIPNK-UHFFFAOYSA-H
gadolinium(3+);trisulfate;octahydrate
16211473
99.9%
Odorless
(REO)
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

missing translation for 'einecsNumber' : 237-115-2

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only