missing translation for 'onlineSavingsMsg'
Learn More

Gadolinium silicide, 99.9% (metals basis), Thermo Scientific™

Chemical Identifiers

CAS 12134-75-7
Molecular Formula GdH8Si2+3
Molecular Weight (g/mol) 221.484
MDL Number MFCD00144382
InChI Key AOLGUBMBPNHJFD-UHFFFAOYSA-N
PubChem CID 71434213
IUPAC Name gadolinium(3+);silane
SMILES [SiH4].[SiH4].[Gd+3]
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11333448
View Documents Promotion Details
Thermo Scientific Alfa Aesar
036356.04
2 g N/A
N/A
11343448
View Documents Promotion Details
Thermo Scientific Alfa Aesar
036356.09
10 g N/A
N/A
Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

12134-75-7
221.484
AOLGUBMBPNHJFD-UHFFFAOYSA-N
gadolinium(3+);silane
GdH8Si2+3
MFCD00144382
71434213
[SiH4].[SiH4].[Gd+3]

Specifications

12134-75-7
GdH8Si2+3
AOLGUBMBPNHJFD-UHFFFAOYSA-N
gadolinium(3+);silane
71434213
99.9%
(metals basis)
Pieces
MFCD00144382
[SiH4].[SiH4].[Gd+3]
221.484
213.42
Odorless
Gadolinium silicide
Videos
Safety and Handling

Safety and Handling

missing translation for 'einecsNumber' : 235-216-6

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications