Learn More
Tocris Bioscience™ Galiellalactone
STAT3 inhibitor; active in vivo
Brand:  Tocris Bioscience™ 6218/500U
245.35 EUR valid until 2024-12-16
Use promo code "21630" to get your promotional price.
Product Code. 18751368
  • 310.00 € / 500µg
Explore more special offers
Description
Description
Galiellalactone is a STAT3 inhibitor. Irreversibly binds STAT3 and inhibits binding to DNA. Inhibits IL-6-dependent JAK/STAT signaling. Induces cell-cycle arrest in G2/M phase and apoptosis in androgen-insensitive prostate cancer cells via activation of ATM/ATR pathway. Also destabilizes microtubules and inhibits cell migration. Exhibits anticancer effects in a mouse tumor xenograft model.
Specifications
Specifications
Inhibitor
133613-71-5
194.23g/mol
STAT Inhibitors
STAT
C11H14O3
Store at -20°C
500 μg