missing translation for 'onlineSavingsMsg'
Learn More

Gallic Acid 98%, Thermo Scientific Chemicals

Product Code. 13498548
Click to view available options
Quantity:
5 g
250 g
1 kg
5 kg
Packaging:
Glass Bottle
Plastic bottle
Plastic drum
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 149-91-7
Molecular Formula C7H6O5
Molecular Weight (g/mol) 170.12
MDL Number MFCD00002510
InChI Key LNTHITQWFMADLM-UHFFFAOYSA-N
Synonym gallic acid, gallate, benzoic acid, 3,4,5-trihydroxy, gallic acid, tech., galop, pyrogallol-5-carboxylic acid, kyselina gallova, 3,4,5-trihydroxybenzoate, ccris 5523, kyselina gallova czech
PubChem CID 370
ChEBI CHEBI:30778
IUPAC Name 3,4,5-trihydroxybenzoic acid
SMILES OC(=O)C1=CC(O)=C(O)C(O)=C1

Specifications

Melting Point 253.0°C
Color Yellow to Beige
Assay Percent Range 98%
Solubility Information Solubility in water: 12g/L cold water. Other solubilities: easily soluble in warmed water, very soluble in ethanol: 160g/L, acetone: 200g/l, soluble in glycerol: 100g/L, slightly soluble in, ether: 10g/L, insoluble in benzene, chloroform, petrol ether
Formula Weight 170.12
Percent Purity 98%
Density 1.6940g/mL
Quantity 5 kg
Infrared Spectrum Authentic
Packaging Plastic drum
Physical Form Powder
Specific Gravity 1.694
Chemical Name or Material Gallic acid, 98%
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.