Learn More
Gallic acid monohydrate, Thermo Scientific Chemicals
61.20 € - 257.00 €
Chemical Identifiers
CAS | 5995-86-8 |
---|---|
Molecular Formula | C7H5O5 |
Molecular Weight (g/mol) | 169.11 |
MDL Number | MFCD00149098 |
InChI Key | LNTHITQWFMADLM-UHFFFAOYSA-M |
Synonym | gallic acid monohydrate, 3,4,5-trihydroxybenzoic acid hydrate, galop hydrate, 3,4,5-trihydroxybenzoic acid monohydrate, gallicum acidum, benzoic acid, 3,4,5-trihydroxy-, hydrate, gallic acid, monohydrate, gallicacidmonohydrate, gallic acid, acs, acmc-1ary0 |
PubChem CID | 24721416 |
IUPAC Name | 3,4,5-trihydroxybenzoic acid;hydrate |
SMILES | OC1=CC(=CC(O)=C1O)C([O-])=O |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10286270
|
Thermo Scientific Acros
220761000 |
100 g | Plastic bottle |
61.20 €
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10677362
|
Thermo Scientific Acros
220765000 |
500 g | Plastic bottle |
69.80 €
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10259493
|
Thermo Scientific Acros
220760025 |
2.5 kg | Plastic bottle |
257.00 €
2.50kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
5995-86-8 | |
169.11 | |
LNTHITQWFMADLM-UHFFFAOYSA-M | |
24721416 | |
OC1=CC(=CC(O)=C1O)C([O-])=O |
C7H5O5 | |
MFCD00149098 | |
gallic acid monohydrate, 3,4,5-trihydroxybenzoic acid hydrate, galop hydrate, 3,4,5-trihydroxybenzoic acid monohydrate, gallicum acidum, benzoic acid, 3,4,5-trihydroxy-, hydrate, gallic acid, monohydrate, gallicacidmonohydrate, gallic acid, acs, acmc-1ary0 | |
3,4,5-trihydroxybenzoic acid;hydrate |
Specifications
5995-86-8 | |
White to Beige | |
(HO)3C6H2CO2H·H2O | |
10, 470 | |
gallic acid monohydrate, 3,4,5-trihydroxybenzoic acid hydrate, galop hydrate, 3,4,5-trihydroxybenzoic acid monohydrate, gallicum acidum, benzoic acid, 3,4,5-trihydroxy-, hydrate, gallic acid, monohydrate, gallicacidmonohydrate, gallic acid, acs, acmc-1ary0 | |
LNTHITQWFMADLM-UHFFFAOYSA-M | |
3,4,5-trihydroxybenzoic acid;hydrate | |
24721416 | |
≥99% (on dry substance) | |
10% max. (120°C) | |
Gallic acid monohydrate, c.p. |
251.0°C | |
C7H5O5 | |
MFCD00149098 | |
15, 4375 | |
Solubility in water: 15g/L (20°C). Other solubilities: soluble in alcohol, glycerol and acetone, practically insoluble in benzene, chloroform and, petroleum ether | |
OC1=CC(=CC(O)=C1O)C([O-])=O | |
169.11 | |
188.12 | |
Authentic | |
Powder |
Safety and Handling
GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Warning
RUO – Research Use Only