missing translation for 'onlineSavingsMsg'
Learn More

Gallium, 99.9999%, (trace metal basis)

Product Code. 10246920 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Plastic bottle
Unit Size:
1g
25g
5g
This item is not returnable. View return policy

Product Code. 10246920

Brand: Thermo Scientific Acros undefined

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 7440-55-3 | Ga | 69.72 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For ultra trace analysis
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7440-55-3
Molecular Formula Ga
Molecular Weight (g/mol) 69.72
MDL Number MFCD00134045
InChI Key GYHNNYVSQQEPJS-UHFFFAOYSA-N
Synonym gallium, elemental, metal, unii-ch46oc8yv4, ch46oc8yv4, ingot, pellets, 6mm dia, galio, metal, packaged in poylethylene bottle, gallium, ion ga1+, atom
PubChem CID 5360835
ChEBI CHEBI:49631
IUPAC Name gallium
SMILES [Ga]

Specifications

Color Gray
Melting Point 29.7°C
Boiling Point 2400.0°C
Density 5.9000g/mL
Quantity 5 g
Packaging Plastic bottle
Assay Percent Range 99.9999%
Form Solid and/or Liquid
Linear Formula Ga
Solubility Information Solubility in water: insoluble.
Formula Weight 69.72
Percent Purity 99.9999%
Grade Trace Metal Basis
Chemical Name or Material Gallium
Show More Show Less
compliance-icons
Product Identifier
  • Gallium
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Substances and mixtures corrosive to metals Category 1
  • Skin corrosion/irritation Category 1 C
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H290-May be corrosive to metals.
Precautionary Statement
  • P234-Keep only in original container.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.