Learn More
Gallium antimonide, 99.99% (metals basis), Thermo Scientific Chemicals
23.00 € - 290.00 €
Chemical Identifiers
CAS | 12064-03-8 |
---|---|
MDL Number | MFCD00016101 |
Synonym | gallium antimonide, stibinogallane, galliumantimonid, gallium antimonide, polycryst, gallium antimonide, single crystal substrate , <100>, diam. x thickness 2 in. x 0.5 mm |
Produktcode | Marke | Quantity | Preis | Menge & Verfügbarkeit | |||||
---|---|---|---|---|---|---|---|---|---|
Produktcode | Marke | Quantity | Preis | Menge & Verfügbarkeit | |||||
11302028
|
Thermo Scientific Alfa Aesar
012933.77 |
0.1 g | |||||||
11322028
|
Thermo Scientific Alfa Aesar
012933.03 |
1 g |
72.40 €
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11312028
|
Thermo Scientific Alfa Aesar
012933.06 |
5 g |
290.00 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Beschreibung
Gallium antimonide can be used for Infrared detectors, infrared LEDs and lasers and transistors, and thermophotovoltaic systems. It can be used as a component of photoresists and other composites where high refractive index is desirable
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemische Identifikatoren
12064-03-8 | |
MFCD00016101 | |
gallium antimonide, stibinogallane, galliumantimonid, gallium antimonide, polycryst, gallium antimonide, single crystal substrate , <100>, diam. x thickness 2 in. x 0.5 mm |
Spezifikation
710°C | |
Pieces | |
UN1549 | |
Insoluble in water. | |
99.99% | |
(metals basis) |
12064-03-8 | |
MFCD00016101 | |
gallium antimonide, stibinogallane, galliumantimonid, gallium antimonide, polycryst, gallium antimonide, single crystal substrate , <100>, diam. x thickness 2 in. x 0.5 mm | |
191.47 | |
Odorless | |
Gallium antimonide |
Sicherheit und Handhabung
GHS H Statement
H302-H332
Harmful if swallowed.
Harmful if inhaled.
P261-P264b-P270-P271-P301+P312-P304+P340-P312-P330-P501c
H302+H332
missing translation for 'dotInformation' : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: ANTIMONY COMPOUNDS, INORGANIC, SOLID, N.O.S.
missing translation for 'einecsNumber' : 235-058-8
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only