missing translation for 'onlineSavingsMsg'
Learn More

Gallium arsenide, 99.999% (metals basis), Thermo Scientific Chemicals

Product Code. 11359269
Click to view available options
Quantity:
2 g
10 g
50 g
This item is not returnable. View return policy
This item is not returnable. View return policy

For Semiconductors (Transistors, Lasers, Solar Cells)

Semiconductors (transistors, lasers, solar cells)Gallium arsenide is used in semiconductor applications. It is also used in the manufacture of devices such as microwave frequency integrated circuits, Gunn diodes, monolithic microwave integrated circuits, infrared light-emitting diodes, laser diodes and optical windows. Further, it is used for single-crystalline thin film solar cells and multi-junction solar cells. In addition, it is used for the detection of X-rays.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1303-00-0
Molecular Formula AsGa
Molecular Weight (g/mol) 144.645
MDL Number MFCD00011017
InChI Key JBRZTFJDHDCESZ-UHFFFAOYSA-N
Synonym gallium arsenide, gallium arsenide gaas, gallium monoarsenide, gallium monoarsenide gaas, unii-27fc46ga44, ccris 4020, arsinogallane, gallium arsenide trace metals basis 1g, gallium arsenide, pieces trace metals basis, gallium arsenide optical window, 12.7mm 0.5in dia x 1mm 0.04in thick
PubChem CID 14770
IUPAC Name gallanylidynearsane
SMILES [Ga]#[As]

Specifications

Melting Point 1238°C
Color Gray
Physical Form Polycrystalline Pieces
Quantity 10 g
Assay Percent Range 99.999% (Metals basis)
Solubility Information Soluble in hydrochloric acid. Insoluble in water,ethanol,methanol and acetone.
Formula Weight 144.64
Odor Odorless
Density 5.31
Chemical Name or Material Gallium arsenide

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'