missing translation for 'onlineSavingsMsg'
Learn More
Gallium(II) chloride, ultra dry, 99.999% (metals basis), Thermo Scientific Chemicals Product Code.: 11323887

Gallium(II) chloride, ultra dry, 99.999% (metals basis), Thermo Scientific Chemicals

Product Code. 11323887
1 g, Ampule
Click to view available options
Quantity:
1 g
Packaging:
Ampule
This item is not returnable. View return policy

Product Code. 11323887

Brand: Thermo Scientific Alfa Aesar 035696.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

 

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Ampouled under argon
TRUSTED_SUSTAINABILITY

Specifications

Boiling Point 535°C
Melting Point 164°C
CAS 24597-12-4
Color White
Physical Form Powder
Quantity 1 g
MDL Number MFCD00151526
UN Number UN3260
Formula Weight 281.26
Percent Purity 99.999%
Odor Acidic
Packaging Ampule
Sensitivity Air and moisture sensitive
Assay (metals basis)
Chemical Name or Material Gallium(II) chloride, Ultra dry
Show More Show Less
compliance-icons
Product Identifier
  • Gallium(II) chloride, ultra dry
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contains : Gallium(II) chloride, ultra dry

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Gallium(II) chloride, ultra dry, 99.999% (metals basis), Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.