Learn More
Gallium(III) 2,4-pentanedionate, 99.99% (metals basis), Thermo Scientific Chemicals
111.00 € - 413.00 €
Chemical Identifiers
CAS | 14405-43-7 |
---|---|
Molecular Formula | C15H21GaO6 |
Molecular Weight (g/mol) | 367.05 |
MDL Number | MFCD00013492 |
InChI Key | IMMRYVXLCAMRNK-UHFFFAOYSA-N |
Synonym | gallium iii acetylacetonate, gallium iii 2,4-pentanedionate |
PubChem CID | 71311502 |
IUPAC Name | (Z)-4-[[(Z)-4-oxopent-2-en-2-yl]oxy-[(E)-4-oxopent-2-en-2-yl]oxygallanyl]oxypent-3-en-2-one |
SMILES | [Ga+3].CC(=O)[CH-]C(C)=O.CC(=O)[CH-]C(C)=O.CC(=O)[CH-]C(C)=O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11379727
|
Thermo Scientific Alfa Aesar
042086.06 |
5 g |
111.00 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11369727
|
Thermo Scientific Alfa Aesar
042086.14 |
25 g |
413.00 €
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
It is used in research on Ga-containing materials. Gallium oxide thin films can be created with atomic layer epitaxy (ALE) by combining gallium acetylacetonate with either water or ozone as the precursor. It can also be used for the synthesis of high purity gallium nitride nano-wires and nano-needles at low temperatures.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
14405-43-7 | |
367.05 | |
IMMRYVXLCAMRNK-UHFFFAOYSA-N | |
71311502 | |
[Ga+3].CC(=O)[CH-]C(C)=O.CC(=O)[CH-]C(C)=O.CC(=O)[CH-]C(C)=O |
C15H21GaO6 | |
MFCD00013492 | |
gallium iii acetylacetonate, gallium iii 2,4-pentanedionate | |
(Z)-4-[[(Z)-4-oxopent-2-en-2-yl]oxy-[(E)-4-oxopent-2-en-2-yl]oxygallanyl]oxypent-3-en-2-one |
Specifications
196°C to 198°C (decomposition) | |
Powder | |
MFCD00013492 | |
Insoluble in water. | |
[Ga+3].CC(=O)[CH-]C(C)=O.CC(=O)[CH-]C(C)=O.CC(=O)[CH-]C(C)=O | |
367.05 | |
367.05 | |
Odorless | |
(metals basis) |
14405-43-7 | |
C15H21GaO6 | |
gallium iii acetylacetonate, gallium iii 2,4-pentanedionate | |
IMMRYVXLCAMRNK-UHFFFAOYSA-N | |
(Z)-4-[[(Z)-4-oxopent-2-en-2-yl]oxy-[(E)-4-oxopent-2-en-2-yl]oxygallanyl]oxypent-3-en-2-one | |
71311502 | |
99.99% | |
1.42 g/mL | |
Gallium(III) 2,4-pentanedionate |
Safety and Handling
P201-P202-P261-P264b-P270-P271-P280i-P281-P301+P312-P302+P352-P304+P340-P305+P351+P338-P308+P313-P312-P330-P332+P313-P362-P501c
H302+H312+H332-H315-H319-H335-H351
missing translation for 'einecsNumber' : 238-377-0
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only