missing translation for 'onlineSavingsMsg'
Learn More

Gallium(III) bromide, anhydrous, Thermo Scientific Chemicals

110.00 € - 366.00 €

Chemical Identifiers

CAS 13450-88-9
Molecular Formula Br3Ga
Molecular Weight (g/mol) 309.435
MDL Number MFCD00016100
InChI Key SRVXDMYFQIODQI-UHFFFAOYSA-K
Synonym gallium tribromide, gallium bromide gabr3, gallium iii bromide, gallium bromide, gabr3, gallium bromide, gabr3, gallium iii bromide, anhydrous, gallium iii bromide, ultra dry, gallium iii bromide, anhydrous, granular
PubChem CID 83477
IUPAC Name tribromogallane
SMILES [Ga](Br)(Br)Br
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11358327
View Documents Promotion Details
Thermo Scientific Alfa Aesar
032107.04
2 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
11368327
View Documents Promotion Details
Thermo Scientific Alfa Aesar
032107.09
10 g
366.00 €
10g
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Gallium(III) bromide is used as organic intermediates. It is a powerful oxidizing agent and is able to release oxygen free radicals from the water in mucous membranes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

13450-88-9
309.435
SRVXDMYFQIODQI-UHFFFAOYSA-K
83477
[Ga](Br)(Br)Br
Br3Ga
MFCD00016100
gallium tribromide, gallium bromide gabr3, gallium iii bromide, gallium bromide, gabr3, gallium bromide, gabr3, gallium iii bromide, anhydrous, gallium iii bromide, ultra dry, gallium iii bromide, anhydrous, granular
tribromogallane

Specifications

279°C
13450-88-9
Crystalline
MFCD00016100
gallium tribromide, gallium bromide gabr3, gallium iii bromide, gallium bromide, gabr3, gallium bromide, gabr3, gallium iii bromide, anhydrous, gallium iii bromide, ultra dry, gallium iii bromide, anhydrous, granular
SRVXDMYFQIODQI-UHFFFAOYSA-K
tribromogallane
83477
Odorless
3.69 g/mL
122°C
White
Br3Ga
UN3260
Slightly soluble in water. Soluble in acids.
[Ga](Br)(Br)Br
309.435
309.45
Moisture sensitive
Gallium(III) bromide, Anhydrous
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c

H314

missing translation for 'dotInformation' : Hazard Class: 8; Packaging Group: III

missing translation for 'einecsNumber' : 236-609-5

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only