missing translation for 'onlineSavingsMsg'
Learn More

Gallium(III) chloride, ultra dry, 99.999% (metals basis), Thermo Scientific Chemicals

89.34 € - 750.74 €

Chemical Identifiers

CAS 13450-90-3
Molecular Formula Cl3Ga
Molecular Weight (g/mol) 176.073
MDL Number MFCD00011018
InChI Key UPWPDUACHOATKO-UHFFFAOYSA-K
Synonym gallium trichloride, gallium iii chloride, trichlorogallium, gallium chloride, gallium chloride gacl3, gallium 3+ chloride, gacl3, ccris 4492, gallium iii chloride, ultra dry, trichlorogalliue
PubChem CID 26010
IUPAC Name trichlorogallane
SMILES Cl[Ga](Cl)Cl
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11330558
SDS View Documents
Thermo Scientific Alfa Aesar
043879.06
5 g
89.34 €
5g
Please to purchase this item. Need a web account? Register with us today!
11354908
SDS View Documents
Thermo Scientific Alfa Aesar
043879.14
25 g
251.25 €
25g
Please to purchase this item. Need a web account? Register with us today!
11340558
SDS View Documents
Thermo Scientific Alfa Aesar
043879.22
100 g
750.74 €
100g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Gallium(III) chloride is used as lewis acid or precursor of organogallium reagents. It is used in detection of solar neutrinos. It is used in semiconductor devices for microelectronics and optics. It acts as a catalyst for a number of transformations such as the ring opening and recyclization of epoxide or trimerization of alkynes and as diamagnetic analogue for ferric compounds.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Ampouled under argon
Specifications

Chemical Identifiers

13450-90-3
176.073
UPWPDUACHOATKO-UHFFFAOYSA-K
26010
Cl[Ga](Cl)Cl
Cl3Ga
MFCD00011018
gallium trichloride, gallium iii chloride, trichlorogallium, gallium chloride, gallium chloride gacl3, gallium 3+ chloride, gacl3, ccris 4492, gallium iii chloride, ultra dry, trichlorogalliue
trichlorogallane

Specifications

201°C
13450-90-3
Crystalline
MFCD00011018
14,4348
Soluble in water,benzene,carbon tetrachloride and carbon disulphide.
Cl[Ga](Cl)Cl
176.073
176.03
Ampule
2.47 g/mL
Gallium (III) chloride, Ultra dry
78°C
White
Cl3Ga
UN3260
gallium trichloride, gallium iii chloride, trichlorogallium, gallium chloride, gallium chloride gacl3, gallium 3+ chloride, gacl3, ccris 4492, gallium iii chloride, ultra dry, trichlorogalliue
UPWPDUACHOATKO-UHFFFAOYSA-K
trichlorogallane
26010
99.999%
Moisture sensitive
(metals basis)
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c

H314-H335

missing translation for 'dotInformation' : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, INORGANIC, N.O.S.

missing translation for 'einecsNumber' : 236-610-0

missing translation for 'rtecsNumber' : LW9100000

missing translation for 'tsca' : Yes

missing translation for 'storageNote' :

  • Ampouled under argon

Recommended Storage : Ambient temperatures; Store under Argon

Product Identifier
  • Gallium(III) chloride, ultra dry

Signal Word
  • Danger

Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B

Hazard Statement
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
  • EUH014-Reacts violently with water.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

Supplemental information
  • MIXTURE LIST-Contains : Gallium trichloride

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.