missing translation for 'onlineSavingsMsg'
Learn More
Gallium(III) nitrate solution, Ga 9-10% w/w, Thermo Scientific Chemicals Product Code.: 11345807

Gallium(III) nitrate solution, Ga 9-10% w/w, Thermo Scientific Chemicals

Product Code. 11345807
250 g
Click to view available options
Quantity:
50 g
250 g
This item is not returnable. View return policy

Product Code. 11345807

Brand: Thermo Scientific Alfa Aesar 043484.30

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Gallium(III) nitrate solution, is used as the material for producing gallium salts and for scientific research and chemical reagent. It is also used as an intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13494-90-1
Molecular Formula GaN3O9
Molecular Weight (g/mol) 255.735
MDL Number MFCD00011016
InChI Key CHPZKNULDCNCBW-UHFFFAOYSA-N
Synonym gallium nitrate, ganite, gallium nitrate anhydrous, nitric acid, gallium salt, unii-y2v2r4w9tq, gallium iii nitrate, ccris 4493, nitric acid, gallium 3+ salt, y2v2r4w9tq, gallium salt
PubChem CID 61635
IUPAC Name gallium;trinitrate
SMILES [N+](=O)([O-])[O-].[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].[Ga+3]

Specifications

Physical Form Liquid
Quantity 250 g
UN Number UN3218
Solubility Information It is soluble in water.
Formula Weight 255.73
Concentration or Composition (by Analyte or Components) Ga 9 to 10% w/w
Chemical Name or Material Gallium (III) nitrate solution

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberM15H018 Certificate TypeCertificate of Analysis Date11/11/2022 Catalog Number043484.30,043484.18
Lot NumberR28G059 Certificate TypeCertificate of Analysis Date18/11/2021 Catalog Number043484.30,043484.18
Lot NumberY19B055 Certificate TypeCertificate of Analysis Date18/11/2021 Catalog Number043484.30,043484.18
Lot NumberD15W025 Certificate TypeCertificate of Analysis Date18/11/2021 Catalog Number043484.30,043484.18
Lot NumberA28M23 Certificate TypeCertificate of Analysis Date18/11/2021 Catalog Number043484.30,043484.18

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Gallium(III) nitrate solution, Ga 9-10% w/w, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.