missing translation for 'onlineSavingsMsg'
Learn More

Gallium(III) nitride, 99.99% (metals basis), Thermo Scientific Chemicals

Product Code. 11399367
Click to view available options
Quantity:
10 g
50 g

Gallium(III) nitride is used to read blu-ray discs. It is used in various wireless infrastructure applications. It finds applications in optoelectronic, high-power and high-frequency devices. It is also useful in military electronics (active electronically scanned array radars). It is biocompatible and used in electrodes and electronic components in living organisms. It is a suitable material for solar cell arrays for satellites. It is employed in bright light-emitting diodes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 25617-97-4
Molecular Formula GaN
Molecular Weight (g/mol) 83.73
MDL Number MFCD00016108
InChI Key JMASRVWKEDWRBT-UHFFFAOYSA-N
Synonym gallium nitride, gallium mononitride, unii-1r9cc3p9vl, gallium nitride,, 1r9cc3p9vl, gallium nitride trace metals basis, gallium iii nitride trace metals basis 10g
PubChem CID 117559
IUPAC Name azanylidynegallane
SMILES N#[Ga]

Specifications

Melting Point 800°C (sublimation)
Color Yellow
Physical Form Powder
Quantity 10 g
Merck Index 14,4351
Solubility Information Slightly soluble in hot concentrated sulfuric acid and hot conc; sodium hydroxide; Insoluble in water and dilute acids.
Formula Weight 83.73
Percent Purity 99.99%
Sensitivity Moisture sensitive
Density 6.1 g/mL
Assay (metals basis)
Chemical Name or Material Gallium(III) nitride
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.