missing translation for 'onlineSavingsMsg'
Learn More

Gallium(III) sulfate hydrate, Puratronic™, 99.999% (metals basis), Thermo Scientific Chemicals

198.00 € - 720.00 €

Chemical Identifiers

CAS 13780-42-2
Molecular Formula Ga2O12S3
Molecular Weight (g/mol) 427.61
MDL Number MFCD00149725
InChI Key UXLRWZBQRAWBQA-UHFFFAOYSA-H
Synonym gallium iii sulfate hydrate, gallium sulphate hydrate, 2ga.3so4.h2o, digallium 3+ hydrate trisulfate, gallium iii sulfate hydrate-ga puratrem, sulfuric acid, galliumsalt 3:2 , hydrate 8ci,9ci
PubChem CID 16213072
IUPAC Name digallium;trisulfate;hydrate
SMILES [Ga+3].[Ga+3].[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11316178
View Documents Promotion Details
Thermo Scientific Alfa Aesar
011152.09
10 g
198.00 €
10g
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
11326178
View Documents Promotion Details
Thermo Scientific Alfa Aesar
011152.18
50 g
720.00 €
50g
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Gallium(III) sulfate hydrate is used in the preparation of other gallium compounds such as gallium oxide. It is supported on silica gel act as a solid acid catalyst, which is employed as a heterogeneous catalyst for the esterification of p-hydroxybenzoic acid and heptanol to give n-heptyl p-hydroxybenzoate. On reacting with strong acid cation ion exchange resin, it exhibits excellent catalytic activity, which is used for synthesizing amyl hexanoate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

13780-42-2
427.61
UXLRWZBQRAWBQA-UHFFFAOYSA-H
16213072
[Ga+3].[Ga+3].[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O
Ga2O12S3
MFCD00149725
gallium iii sulfate hydrate, gallium sulphate hydrate, 2ga.3so4.h2o, digallium 3+ hydrate trisulfate, gallium iii sulfate hydrate-ga puratrem, sulfuric acid, galliumsalt 3:2 , hydrate 8ci,9ci
digallium;trisulfate;hydrate

Specifications

105°C to 110°C
Crystalline Aggregates
MFCD00149725
Slightly soluble in water and acid.
[Ga+3].[Ga+3].[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O
427.61
427.62 (Anhydrous)
Puratronic™
Hygroscopic
(metals basis)
13780-42-2
Ga2O12S3
gallium iii sulfate hydrate, gallium sulphate hydrate, 2ga.3so4.h2o, digallium 3+ hydrate trisulfate, gallium iii sulfate hydrate-ga puratrem, sulfuric acid, galliumsalt 3:2 , hydrate 8ci,9ci
UXLRWZBQRAWBQA-UHFFFAOYSA-H
digallium;trisulfate;hydrate
16213072
99.999%
Odorless
3.860 g/mL
Gallium(III) sulfate hydrate
Videos
Safety and Handling

Safety and Handling

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only