missing translation for 'onlineSavingsMsg'
Learn More

Gallium Indium alloy, 99.99% (metals basis), Thermo Scientific Chemicals

38.59 € - 659.28 €

Specifications

Chemical Name or Material Gallium Indium alloy
Color Gray
Assay Percent Range 99.99% (metals basis)
Molecular Formula Ga:In; 40:60 wt.%
MDL Number MFCD00144387
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11339768
View Documents
Thermo Scientific Alfa Aesar
044240.03
1 g
38.59 €
1g
Please to purchase this item. Need a web account? Register with us today!
11319768
View Documents
Thermo Scientific Alfa Aesar
044240.09
10 g
164.82 €
10g
Please to purchase this item. Need a web account? Register with us today!
11329768
View Documents
Thermo Scientific Alfa Aesar
044240.18
50 g
659.28 €
50g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Step soldering involving temperature sensitive components, Soldering to molded interconnect device (MID) plastics, Fusible alloys/fuse applications, Mercury replacement, Thermal and electrical conductivity.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Gallium Indium alloy
99.99% (metals basis)
MFCD00144387
Odorless
Gray
Ga:In; 40:60 wt.%
Insoluble in water.
Solid
Videos
Safety and Handling

Safety and Handling

P234-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P390-P501c

H290-H314-H335

missing translation for 'einecsNumber' : 231-163-8

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.