missing translation for 'onlineSavingsMsg'
Learn More

Gallium Indium eutectic, 99.99% (metals basis), Thermo Scientific Chemicals

Gallium Indium eutectic, Ga:In; 75.5:24.5 wt%, 50 g, 99.99% (metals basis), 6.2, Hazard Class: 8; Packaging Group: III, 231-163-8, H290-H314-H335, P234-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P390-P501c, 15.7 deg.C, MFCD00144387, Liquid

40.20 € - 622.10 €

Specifications

Chemical Name or Material Gallium Indium eutectic
Melting Point 15.7°C
Density 6.2
Assay Percent Range 99.99% (metals basis)
Molecular Formula Ga:In; 75.5:24.5 wt%
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11341819
View Documents
Thermo Scientific Alfa Aesar
012478.03
1 g
40.20 €
1g
Please to purchase this item. Need a web account? Register with us today!
11331819
View Documents
Thermo Scientific Alfa Aesar
012478.09
10 g
173.87 €
10g
Please to purchase this item. Need a web account? Register with us today!
11351819
View Documents
Thermo Scientific Alfa Aesar
012478.18
50 g
622.10 €
50g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

It useful as a contact electrode for the electrical characterization of thin film organic and semiconductor devices. Gallium-Indium eutectic top contact, and a halogen light source was used to test P3HT-PCBM organic solar cells.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Gallium Indium eutectic
6.2
Ga:In; 75.5:24.5 wt%
Insoluble in water or organic solvents.
15.7°C
99.99% (metals basis)
MFCD00144387
Liquid
Videos
Safety and Handling

Safety and Handling

P234-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P390-P501c

H290-H314-H335

missing translation for 'dotInformation' : Hazard Class: 8; Packaging Group: III

missing translation for 'einecsNumber' : 231-163-8

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only