missing translation for 'onlineSavingsMsg'
Learn More

Gallium ingot, 99.99% (metals basis), Thermo Scientific Chemicals

223.11 € - 1634.13 €

Chemical Identifiers

CAS 7440-55-3
Molecular Formula Ga
Molecular Weight (g/mol) 69.72
MDL Number MFCD00134045
InChI Key GYHNNYVSQQEPJS-UHFFFAOYSA-N
Synonym gallium, elemental, metal, unii-ch46oc8yv4, ch46oc8yv4, ingot, pellets, 6mm dia, galio, metal, packaged in poylethylene bottle, gallium, ion ga1+, atom
PubChem CID 5360835
ChEBI CHEBI:49631
IUPAC Name gallium
SMILES [Ga]
View More Specs

Products 4
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11305759
View Documents
Thermo Scientific Alfa Aesar
010188.14
25 g
223.11 €
25g
Please to purchase this item. Need a web account? Register with us today!
11391238
View Documents
Thermo Scientific Alfa Aesar
010188.18
50 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
11301248
View Documents
Thermo Scientific Alfa Aesar
010188.30
250 g
1044.20 €
250g
Please to purchase this item. Need a web account? Register with us today!
11381238
View Documents
Thermo Scientific Alfa Aesar
010188.36
500 g
1634.13 €
500g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

7440-55-3
69.72
GYHNNYVSQQEPJS-UHFFFAOYSA-N
5360835
gallium
Ga
MFCD00134045
gallium, elemental, metal, unii-ch46oc8yv4, ch46oc8yv4, ingot, pellets, 6mm dia, galio, metal, packaged in poylethylene bottle, gallium, ion ga1+, atom
CHEBI:49631
[Ga]

Specifications

Gray-Silver
Ga
UN2803
GYHNNYVSQQEPJS-UHFFFAOYSA-N
gallium
5360835
99.99%
(metals basis)
7440-55-3
MFCD00134045
gallium, elemental, metal, unii-ch46oc8yv4, ch46oc8yv4, ingot, pellets, 6mm dia, galio, metal, packaged in poylethylene bottle, gallium, ion ga1+, atom
[Ga]
69.72
CHEBI:49631
Ingot
Gallium ingot
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H290
May be corrosive to metals.

P234-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P390-P501c

H290-H314-H335

missing translation for 'dotInformation' : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: GALLIUM

missing translation for 'einecsNumber' : 231-163-8

missing translation for 'rtecsNumber' : LW8600000

missing translation for 'tsca' : Yes

missing translation for 'storageNote' : Requires overnight shipping with cold pack; Store at 2°C to 8°C

Recommended Storage : Keep cold

Product Identifier
  • Gallium ingot

Signal Word
  • Warning

Hazard Category
  • Substances and mixtures corrosive to metals Category 1

Hazard Statement
  • H290-May be corrosive to metals.

Precautionary Statement
  • P234-Keep only in original container.
  • P390-Absorb spillage to prevent material damage.
  • P406-Store in corrosive resistant/ container with a resistant inner liner.

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.