missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gallium ingot, 99.99% (metals basis), Thermo Scientific Chemicals
missing translation for 'orderingAttributeHoverText'
Quantity:
25 g
50 g
250 g
500 g
missing translation for 'unitSize'
250g
25g
500g
50g
Beschreibung
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Spezifikation
Spezifikation
| Color | Silver to Gray |
| Appearance | Silver Gray ingot |
| Quantity | 250 g |
| Assay Percent Range | 99.9999% (Metals basis) |
| Form | Ingot |
| Chemical Name or Material | Gallium |
RUO – Research Use Only
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur