missing translation for 'onlineSavingsMsg'
Learn More

Gallium ingot, 99.99% (metals basis), Thermo Scientific Chemicals

Product Code. 11385749
Click to view available options
Quantity:
25 g
50 g
250 g
500 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Requires overnight shipping with cold pack

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7440-55-3
Molecular Formula Ga
Molecular Weight (g/mol) 69.72
MDL Number MFCD00134045
InChI Key GYHNNYVSQQEPJS-UHFFFAOYSA-N
Synonym gallium, elemental, metal, unii-ch46oc8yv4, ch46oc8yv4, ingot, pellets, 6mm dia, galio, metal, packaged in poylethylene bottle, gallium, ion ga1+, atom
PubChem CID 5360835
ChEBI CHEBI:49631
IUPAC Name gallium
SMILES [Ga]

Specifications

Color Silver to Gray
Appearance Silver Gray ingot
Assay Percent Range 99.9999% (Metals basis)
Quantity 25 g
Form Ingot
Chemical Name or Material Gallium
compliance-icons
Product Identifier
  • Gallium ingot
Signal Word
  • Warning
Hazard Category
  • Substances and mixtures corrosive to metals Category 1
Hazard Statement
  • H290-May be corrosive to metals.
Precautionary Statement
  • P234-Keep only in original container.
  • P390-Absorb spillage to prevent material damage.
  • P406-Store in corrosive resistant/ container with a resistant inner liner.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'