missing translation for 'onlineSavingsMsg'
Learn More
Learn More
WTW™ Galvanic Dissolved Oxygen Sensors (universal)
Get accurate testing results for water and waterous solutions with these membrane-covered galvanic dissolved oxygen sensors that are low maintenance and no polarizarion time required.
Brand: WTW™ 201545
1098.00 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
This item is not returnable.
View return policy
Description
- Dissolved oxygen sensor with integrated temperature compensation and membrane leak monitoring
- Can be used in the lab as well as in the field
- Features include a quick response time (t99<60 s) and low-maintenance operation
- Fields of use include surface waters, measurements in wastewater treatment plants, BOD directions
- Filled with electrolyte and achieves a long dwelling time up to 6 months
- Calibration vessel is part of the delivery
- For use with conductivity and multiparameter meters
- Includes 1.5m L Cable Cleaning solution, abrasive foil, 3 exchange membrane heads and electrolyte
Specifications
3 m | |
3m L Cable Cleaning solution, abrasive foil, 3 exchange membrane heads and electrolyte | |
Conductivity and multiparameter meters |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction