missing translation for 'onlineSavingsMsg'
Learn More

WTW™ Galvanic Dissolved Oxygen Sensor for WTW Conductivity and Multiparameter

proven conventional sensors for pH, ORP, ISE, conductivity and dissolved oxygen are the masters of all measurements in the field and in the lab

1430.00 €

Specifications

Length (Metric) Cable 20 m
Includes 20m L Cable Cleaning solution, abrasive foil, 3 exchange membrane heads and electrolyte
For Use With (Equipment) Conductivity and multiparameter meters
View More Specs

Products 1
Product Code Brand Description Price Quantity & Availability  
Product Code Brand Description Price Quantity & Availability  
10160691
View accessory products View Documents
WTW™
201425
StirrOx™ G Self-stirring Dissolved Oxygen Sensor
1430.00 €
Each
Estimated Shipment: 16-01-2025
to see stock.
Add to basket
 
Description

Description

The CellOx™ 325 is a dissolved oxygen sensor with integrated temperature compensation and membrane leak monitoring, which can be used in the lab as well as in the field. Its features include a quick response time (t99<60 s) and low-maintenance operation. Its fields of use include surface waters, measurements in wastewater treatment plants, BOD directions, but also industrial and other applications. It is filled with electrolyte and achieves a long dwelling time up to 6 months. The calibration vessel is part of the delivery.
Specifications

Specifications

20 m
20m L Cable Cleaning solution, abrasive foil, 3 exchange membrane heads and electrolyte
Conductivity and multiparameter meters
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.