missing translation for 'onlineSavingsMsg'
Learn More

Manutan Galvanised rope grip, 1000kg WLL capacity, 10mm diameter

Brand:  Manutan A341563

Encompass_Preferred

Product Code. 12888688

  • 3.00 € / Each
Estimated Shipment: 27-01-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Galvanised rope grips
  • Diameter: 10mm
  • WLL: 1000kg
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers