missing translation for 'onlineSavingsMsg'
Learn More

Manutan Galvanised rope grip, 700kg WLL capacity, 8mm diameter

Brand:  Manutan A341548

Encompass_Preferred

Product Code. 12878678

  • 3.00 € / Each
Estimated Shipment: 23-01-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Galvanised rope grips
  • Diameter: 8mm
  • WLL: 700kg
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers