missing translation for 'onlineSavingsMsg'
Learn More

Manutan Galvanised thimble, 1400kg WLL capacity, 11mm diameter

Brand:  Manutan A341551

Encompass_Preferred

Product Code. 12858698

  • 3.00 € / Each
Estimated Shipment: 16-04-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Galvanised thimbles
  • Diameter: 11
  • WLL: 1400
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers