missing translation for 'onlineSavingsMsg'
Learn More

Manutan Galvanised thimble, 275kg WLL capacity, 5mm diameter

Brand:  Manutan A339846

Encompass_Preferred

Product Code. 12808668

  • 0.94 € / Each
Estimated Shipment: 16-04-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Galvanised thimbles
  • WLL: 275kg
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers