Learn More
Keystone Cleanroom Products Gamma-Irradiated 2 mil Polyethylene Trash Bag
GREENER_CHOICE
Simplify the removal of waste from your aseptic cleanroom without introducing external sources of microbial contamination, by using gamma irradiated trash liners.
Brand:  Keystone Cleanroom Products S-2428CP-10
 View more versions of this product
Product Code. 17706879
  • 621.00 € / Pack of 10
Estimated Shipment: 09-07-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Packaged in an ISO6 cleanroom
  • 2 mil clear polyethylene
  • Each package includes a lot number for traceability
  • Certificates of Conformance and Irradiation sent with shipment
  • Packaged in Tyvek™/Mylar Chevron Peel Pouches
Specifications
Specifications
Not Autoclavable
Clear
LDPE
2 mil
16.3 L
10 Packs
0.05 mm
71 x 61 cm (28 x 24 in.)