Learn More
Fisherbrand™ Gamma Radiation Safe Block for Eppendorf Tubes
Provides radiation protection shielding for use in laboratories.
Brand:  Fisherbrand™ CSR-BLOCKG
Product Code. 12357137
  • 692.00 € / Each
Estimated Shipment: 07-03-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Transparent shielding
  • Serves as solution for short-term storage of radioactive waste or radioisotopes
  • Constructed of acrylic provides easy fabrication and manufacturability
Specifications
Specifications
3.5 x 14 x 5cm (1.38 x 5.51 x 1.97 in.)
4 x 1.5mL Eppendorf tubes
Acrylic
Gamma Radiation Safe
Block for Eppendorf tubes