Learn More
Fisherbrand™ Gamma Radiation Safe Transport Box
Provides radiation protection shielding for use in laboratories.
Brand:  Fisherbrand™ CSR-B8G
Product Code. 12377127
  • 4800.00 € / Each
Estimated Shipment: 04-04-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Manufactured with hinged lids
  • Transparent shielding
  • Serves as solution for short-term storage of radioactive waste or radioisotopes
  • Constructed of acrylic provides easy fabrication and manufacturability
Specifications
Specifications
29.5 x 38 x 7.5cm (11.61 x 14.96 x 2.95 in.)
Radioisotopes or radioactive waste
Transport Box
Gamma radiation shielded
Hinged lid
Acrylic