missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals gamma-Secretase Inhibitor I

Product Code. 15466179
Click to view available options
Quantity:
5 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

 

Cell permeable. Inhibits --secretase, a protease that cleaves the amyloid precursor protein (APP) to generate --amyloid protein

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

Quantity 5 mg
Description White
Content And Storage -30°C to -10°C
For Use With (Application) Cell permeable. Inhibits →-secretase, a protease that cleaves the amyloid precursor protein (APP) to generate →-amyloid protein
CAS 133407-83-7
Molecular Formula C26H41N3O5
Formula Weight 475.62
Synonym Z-Leu-Leu-Nle-CHO (Nle = Norleucine);
Sensitivity Hygroscopic
Product Type Gamma-Secretase Inhibitor I
Form Solid
InChI Key RNPDUXVFGTULLP-UHFFFAOYNA-N
SMILES CCCCC(NC(=O)C(CC(C)C)NC(=O)C(CC(C)C)NC(=O)OCC1=CC=CC=C1)C=O
IUPAC Name benzyl N-[3-methyl-1-({3-methyl-1-[(1-oxohexan-2-yl)carbamoyl]butyl}carbamoyl)butyl]carbamate
Molecular Weight (g/mol) 475.63
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.