Learn More
Berkshire™ Gamma™ SuperSorb™ Low Endotoxin Wipe
Berkshire’s line of low endotoxin cleanroom wipes have been engineered to meet the specific requirements of the pharmaceutical, healthcare and medical device industries.
Brand: Berkshire™ GWSSST16LE
This item is not returnable.
View return policy
Specifications
Polyester | |
Ultrasonically Sealed | |
guaranteed endotoxin limits - <20 EU/device - <0.5 EU/mL |
Knitted | |
30 x 30 cm |