Learn More
Berkshire™ Gamma Wipe™ 120 Cleanroom Wipers
BIOPHARMA
Gamma Wipe™ 120 is a sterile cleanroom laundered 100% polyester knit wiper.
800.00 €
Specifications
Product Type Cleanroom Wipe
Material Polyester
Absorbancy Highly Absorbent
Compatibility Common cleaning and disinfecting solutions
Autoclavable Not Autoclavable
View More Specs
Products 1
Product Code Brand Dimensions (L x W) Price Quantity & Availability  
Product Code Brand Dimensions (L x W) Price Quantity & Availability  
12887143
BIOPHARMA
View Documents
Berkshire™
GW120ST15
30.48 x 30.48 cm (12 x 12 in.)
800.00 €
Pack of 300
Estimated Shipment: 01-04-2025
to see stock.
Add to basket
Description
Description
  • Low particles, fibers, ions and extractables
  • Continuous-filament polyester knit fabric
  • High absorbency
  • High abrasion resistance
  • Soft hand and edge for sensitive surfaces
  • Chemically compatible with common cleaning and disinfecting solutions including isopropyl alcohol, phenols, bleaches and quarternary ammonium compounds
  • For use in ISO Class 4 (Class 10) applications
Specifications
Specifications
Cleanroom Wipe
Highly Absorbent
Not Autoclavable
Knit
For use in ISO Class 4 and higher sterile environments where microbial exclusion is demanded, excellent for cleaning medical devices
1-ply
Polyester
Common cleaning and disinfecting solutions
RoHS 2002/05, REACH
Knife Cut
For cleaning sterile laminar air flow units, barrier isolators and bio-containment units
Non-sterile