Learn More
GAN Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 14305-1-AP-20UL
 View more versions of this product
Product Code. 16852884
  • 145.00 € / 20µL
Explore more special offers
Description
Description
This gene encodes a member of the cytoskeletal BTB/kelch repeat family. The encoded protein plays a role in neurofilament architecture and is involved in mediating the ubiquitination and degradation of some proteins. Defects in this gene are a cause of giant axonal neuropathy.
Specifications
Specifications
GAN
Polyclonal
Unconjugated
GAN
GAN, GAN1, gigaxonin, Kelch like protein 16, KLHL16
Rabbit
Antigen Affinity Chromatography
RUO
209239, 307893, 8139
-20°C
Liquid
Western Blot
0.28 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q8CA72, Q9H2C0
GAN
GAN Fusion Protein Ag5421
20 μL
Primary
Rat, Mouse, Human