missing translation for 'onlineSavingsMsg'
Learn More

Cole-Parmer™ Gardner Denver™ Vacuum/Pressure Diaphragm Pump

Wetted parts are coated with PTFE to handle your toughest applications

Brand:  Cole-Parmer™ 79200-40

545.30 EUR valid until 2025-02-21
Use promo code "24115" to get your promotional price.


Product Code. 13346259

  • 574.00 € / Each
Estimated Shipment: 11-03-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Lightweight aluminum construction and compact size make pumps ideal for benchtop applications such as vacuum filtration.
  • Pumps with gauges and regulators give you more control of your vacuum/pressure applications.
  • Include suction cup feet and a 6-ft power cord with built-in switch (the 115 VAC models have grounded plug, 220 VAC and DC models have stripped ends).
Specifications

Specifications

178.3 mmHg
Permanent magnet
Port size: 1/8 in. NPT(F)
104°F
33.6 L/min. Max.
18.73 mm
1/10 hp
5 A
PTFE
40°C
120.7 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.