missing translation for 'onlineSavingsMsg'
Learn More

Lenz Laborglasintrumente Gas Sampling Tubes

Made of high quality materials. Each manufacturing step is tested through an extensive quality management system, based on DIN ISO 9001/EN 29001.

Brand:  Lenz Laborglasintrumente 05680070

 View more versions of this product

Product Code. 10201901

  • 130.00 € / Each
Estimated Shipment: 05-06-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Gas sampling tubes acc. to DIN 12473-1
  • Available with or without nozzle (with thread GL14 and septa)
  • With two one-way stopcocks NS 14 (with retaining device)
  • Connections: capillary tube (outer dia. 8 mm, inner dia. 2,7 mm)
  • Glass olive (outer dia. 9 mm)
  • With inscription label
  • Made of DURAN™ tubing
Specifications

Specifications

1000 mL
Duran Glass
Without nozzle
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

One moment while we fetch your results.
Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Lenz Laborglasintrumente Gas Sampling Tubes > 1000 mL

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.