missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BINDER™ O2 Gas Tank Connection Kit
For Use With: series CB
Brand: BINDER™ 8012-0015
This item is not returnable.
View return policy
Description
- Max. pressure 10 bar
- With connection parts
- 5 m hose included
- Union nut and elbow
- Anti-static vacuum hose
- For use in CB series incubators
Specifications
Gas cylinder connection set - for O2, consisting of a gas tank pressure regulator with connection parts and 5-meter hose | |
Gas Tank Connection Kit | |
10 bar |
BINDER Chambers Series CB, CBF | |
Connection parts, 5 m hose |
Product Suggestions
Customers who viewed this item also viewed
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
BINDER™ O2 Gas Tank Connection Kit > For Use With: All CB models
Spot an opportunity for improvement?Share a Content Correction