Learn More
Trajan™ Gas-Tight Autosampler Syringes
Replace syringes for Agilent™ Technologies, Varian™, Perkin Elmer™, Shimadzu™, Carlo Erba™, Fisons™, CTC/Leap™, Hitachi™, and more
Brand:  Trajan™ 005330
 View more versions of this product
Product Code. 12997556
  • 114.00 € / Each
Explore more special offers
Description
Description
Syringe needles are cone point unless otherwise noted.
Order Info
Additional syringes are available for other autosamplers made by the manufacturers represented here, as well as syringes for Unicam™, Waters™, and Siemens™ instruments. For information, contact your Fisher Scientific Customer Service Representative.
Specifications
Specifications
22 G
0.72 mm
2 in.
100R-C/T-GT-LC
100 μL
Carlo Erba/Fisons/Leap/CTC ASD200 and AS800
0.02 in.
High performance liquid chromatology
Autosampler Syringe
Ea.
51 mm
Removable