Learn More
Trajan Scientific and Medical™ Gas Tight Fixed Needle Autosampler Syringe, Cone Tip
Designed to provide improved solvent resistance, greater temperature range and increased operational smoothness.
Brand:  Trajan Scientific and Medical™ 007862
 View more versions of this product
Product Code. 13126105
  • 129.00 € / Each
Explore more special offers
Description
Description
  • High quality solutions for liquid handling
  • Accurate and reproducible injections
  • Specifically designed for Thermo Scientific instruments
Specifications
Specifications
PTFE-tipped (Plunger)
0.32 mm
Gastight
57 mm
Fixed
23 G
0.63 mm
500 mL
Cone