missing translation for 'onlineSavingsMsg'
Learn More

RETSCH Gasket For Grinding Jar

Brand:  RETSCH 03.111.0291

 View more versions of this product

Product Code. 15330984

  • 26.45 € / Each
Estimated Shipment: 09-01-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Specifications

Specifications

For 25mL grinding jar
1 Pack
Gasket
25 mL Grinding Jar
Stainless Steel, PTFE
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers