missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BD GasPak™ 150 Replacement Parts and Accessories
134.00 € - 295.00 €
Product Code | Brand | Type | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Type | Price | Quantity & Availability | ||||||
15991652
|
BD
270124 |
Inner Lid |
134.00 €
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||||
11729022
|
BD
260610 |
Lid Assembly | N/A | |||||||
12340439
|
BD
260607 |
Anaerobic Jar | N/A | |||||||
11739022
|
BD
260618 |
Petri Dish Rack |
240.00 €
Each |
Estimated Shipment: 12-05-2025
Log in to see stock. |
||||||
Description
Specifications
GasPak™ 150 |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- Catalyst Charges (GasPak 150)
Signal Word
- Warning
Hazard Category
- Serious eye damage/eye irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
Precautionary Statement
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P264-Wash thoroughly after handling.
- P271-Use only outdoors or in a well-ventilated area.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P337+P313-If eye irritation persists: Get medical advice/attention.
- P403+P233-Store in a well-ventilated place. Keep container tightly closed.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : alumina
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
BD GasPak™ 150 Replacement Parts and Accessories