Learn More
Thermo Scientific™ Cover for Precision™ Water Baths
Complement your water baths with a gassing hood, gable cover, designed to fit large flasks, or concentric ring cover for steaming.
Brand:  Thermo Scientific™ 3162639
 View more versions of this product
Product Code. 15335897
  • 241.20 € / Each
Estimated Shipment: 06-06-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
Complement your water baths with a gassing hood, gable cover, designed to fit large flasks, or concentric ring cover for steaming.
Gassing Hoods
  • Permit control over the atmosphere surrounding the sample
  • Improve temperature uniformity and reduce energy consumption
  • Polished stainless steel with plastic gas fitting
  • Each bath will accommodate 1 large hood or 2 small hoods
Services
1 year
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Small Gassing Hood
Precision 15L Shaking Water Baths
Gassing Hood