Learn More
Thermo Scientific™ Gateway Accessory Kit for OPC™ UA enablement of Bio Safety Cabinets
Benefit from paperless record keeping, remote monitoring, cloud analytics and more by incorporating your Thermo Scientific™ Biological Safety Cabinet into your Distributed Control Systems (DCS) using the Gateway Accessory Kit for OPC™ UA enablement.
2200.95 €
Specifications
Product Type | Gateway Accessory Kit |
---|---|
Includes | Power supply 100-240 V, USB 2.0 cable USB-A to USB-B connector |
For Use With (Equipment) | Herasafe™ 2030i, Maxisafe 2030i, Herasafe™ 2025, MSC-Advantage™, 1300 Series A2, and 1500 Series A2 Biological Safety Cabinets |
Voltage | 100/240 V |
Product Code | Brand | Description | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Description | Price | Quantity & Availability | |||||
17907202
|
Thermo Scientific™
50170006 |
Thermo Scientific OPC UA Gateway for Bio Safety Cabinets available in all countries except for China |
2200.95 €
1 unit |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Enable Open Platform Communication Unified Architecture (OPC™ UA) integration of Thermo Scientific™ Biological Safety Cabinets using this gateway accessory kit. Benefit from the full potential of incorporating lab equipment into your new or existing Distributed Control Systems (DCS), including paperless record keeping, remote monitoring, cloud analytics and more.
Thermo Scientific™ OPC™ UA Gateway for Biological Safety Cabinets utilizes the OPC™ UA interoperability communication protocol to enable a secure and reliable exchange of real-time and historical data between Distributed Control Systems (DCS), such as the Emerson™ DeltaV™ system, and other systems, applications, and enterprise users.
What is OPC UA?
Open Platform Communication Unified Architecture (OPC™ UA) is a platform-independent, open industry standard for connectivity that enables machine-to-machine communication and information integration between various devices, software systems, and applications.
Advantages to connecting
- 21 CFR Part 11–compliance and GMP needs
- Support audit requirements with enhanced data traceability
- Real-time remote monitoring, workflow efficiencies, and assets and cost optimization, and regulatory compliance.
The gateway accessory kit comes complete with a gateway device, USB 2.0 cable USB-A to USB-B connector, and 100-240 V power supply. One kit is needed per Biological Safety Cabinet connection.
The gateway accessory kit is compatible with the following models:
- Thermo Scientific™ Herasafe™ 2030i Biological Safety Cabinet
- Thermo Scientific™ Maxisafe 2030i Biological Safety Cabinet
- Thermo Scientific™ Herasafe™ 2025 Biological Safety Cabinet
- Thermo Scientific™ MSC-Advantage™ Biological Safety Cabinet
- Thermo Scientific™ 1300 Series A2 Biological Safety Cabinet
- Thermo Scientific™ 1500 Series A2 Biological Safety Cabinet
Specifications
Gateway Accessory Kit | |
Power supply 100-240 V, USB 2.0 cable USB-A to USB-B connector | |
Herasafe™ 2030i, Maxisafe 2030i, Herasafe™ 2025, MSC-Advantage™, 1300 Series A2, and 1500 Series A2 Biological Safety Cabinets | |
100/240 V |