Learn More
Macherey-Nagel™ GC Column Me-Sil Retention Gap
219.00 € - 545.00 €
Product Code | Brand | Diameter (Metric) Inner | Length (Metric) | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Diameter (Metric) Inner | Length (Metric) | Price | Quantity & Availability | ||||
10398541
|
Macherey-Nagel™
723706.10 |
0.25 mm | 10 m |
219.00 €
Each |
Estimated Shipment: 18-11-2024
Log in to see stock. |
||||
10358591
|
Macherey-Nagel™
723706.25 |
0.25 mm | 25 m |
404.00 €
Each |
Estimated Shipment: 18-11-2024
Log in to see stock. |
||||
10429171
|
Macherey-Nagel™
723707.10 |
0.32 mm | 10 m |
247.00 €
Each |
Estimated Shipment: 18-11-2024
Log in to see stock. |
||||
10429361
|
Macherey-Nagel™
723707.25 |
0.32 mm | 25 m |
451.00 €
Each |
Estimated Shipment: 18-11-2024
Log in to see stock. |
||||
10002841
|
Macherey-Nagel™
723708.10 |
0.53 mm | 10 m |
292.00 €
Each |
Estimated Shipment: 18-11-2024
Log in to see stock. |
||||
10236611
|
Macherey-Nagel™
723708.25 |
0.53 mm | 25 m |
545.00 €
Each |
Estimated Shipment: 18-11-2024
Log in to see stock. |
||||
Specifications
GC Column Retention Gap | |
320°C | |
1 Each |