missing translation for 'onlineSavingsMsg'
Learn More

Trajan™ Sealing Ring for GC Ferrule

Graphite sealing ring for use with GC Ferrules

58.40 €

Specifications

Material 100% Graphite
Type Sealing Ring
View More Specs

Products 2
Product Code Brand Diameter (Metric) Inner Length (Metric) Width (Metric) Price Quantity & Availability  
Product Code Brand Diameter (Metric) Inner Length (Metric) Width (Metric) Price Quantity & Availability  
10578102
View Documents
Trajan™
0726003
6.35 mm 2.2 mm 9.9 mm
58.40 €
Each
Estimated Shipment: 23-05-2025
to see stock.
Add to basket
 
15298873
View Documents
Trajan™
0726520
1.59 mm 1.7 mm 3.2 mm
58.40 €
Pack of 10
Please to purchase this item. Need a web account? Register with us today!
Description

Description

  • For use with GC Ferrules
  • 100% Graphite Sealing ring for 1/4 in. fittings
  • Supplied in packs of 10
Specifications

Specifications

100% Graphite
Sealing Ring
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Trajan™ Sealing Ring for GC Ferrule

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.