Learn More
Trajan Scientific and Medical™ GC Inlet Liner Split/Splitless with Recessed Gooseneck Quartz Wool For Shimadzu 17A And 2014 3.4mm I.D.
The unique deactivation process of SGE inlet liners provides excellent analysis for a wide range of compounds
Brand:  Trajan Scientific and Medical™ 09206101
Product Code. 12973991
  • 48.70 € / Each
Estimated Shipment: 29-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
Product
Specifications
Specifications
1 Ea.
GC Inlet Liner, Split / Splitless Inlet Liner with Recessed Gooseneck and Quartz Wool
5 mm
95 mm
3.4 mm
Shimadzu GC Instruments