Learn More
Trajan™ GC Inlet Liners
For use with GC instruments
Brand:  Trajan™ 092144
 View more versions of this product
Product Code. 10085270
  • 268.00 € / Pack of 5
Estimated Shipment: 19-12-2024
to see stock.
Explore more special offers
Add to basket
Description
Description
For use with GC instruments
  • All liners are deactivated using high temperature (420°C [788°F]) gas phase deactivation
  • Supplied in blister packs to prevent contamination
  • Reduced active sites mean no unwanted reaction in samples
FocusLiners™
  • Include two tapered sections that secure the quartz wool plug for optimum needle tip wiping and sample transfer—also prevent quartz wool from shifting towards the septum or pushing down into the liner
  • Provide excellent reproducibility
  • Results in lower mass discrimination
  • Fast FocusLiners have a narrow 2.3mm I.D. for use with 0.1mm I.D. capillary columns—narrow I.D. also results in smaller peak widths
Liners for Split Injection
  • Quartz wool in some split liners promotes analyte mixing and results in better quantitation, provides a large surface area that aids in liquid sample vaporization, acts as a trap to collect nonvolatile residue from sample, and prevents sample from hitting the bottom of the injector before volatilization
  • Liners for Splitless Injection
    • Tapers in some splitless liners minimize contact of analytes with the bottom of the injection port –especially useful for very active compounds; also channel analytes into column for a more robust column installation
    Specifications
    Specifications
    Splitless with single taper
    Inlet Liners
    8 mm
    105 mm
    5 mm