Learn More
Trajan Scientific and Medical™ GC Tubing Fused Silica (Not Deactivated)
Helps in maintaining a high performance chromatography system
Brand:  Trajan Scientific and Medical™ 0624635
 View more versions of this product
Product Code. 15288823
  • 122.00 € / 10m
Estimated Shipment: 23-12-2024
to see stock.
Explore more special offers
Add to basket
Description
Description
  • High quality fused silica tubing
  • Tubing protected with high temperature polyimide resin
  • Used in a wide range of GC, LC and biotechnology applications
Specifications
Specifications
Tubing
0.05 mm
10 m
GC, LC and Biotechnology
0.15 mm