missing translation for 'onlineSavingsMsg'
Learn More

Cytiva Gel Filtration Calibration Kits

Calibrate gel filtration columns simply, reliably. Cytiva Life Sciences™ Gel Filtration Calibration Kits contain a well-defined series of protein standards.

670.00 €

Specifications

Quantity Each
For Use With (Equipment) High-resolution media and prepacked column
Product Type Calibration Kit
For Use With (Application) Calibration of high-resolution gel filtration media and column such as Superdex™, Superose and Sephacryl
Content And Storage 2° to 8°C
View More Specs

Products 2
Product Code Brand MWCO Price Quantity & Availability  
Product Code Brand MWCO Price Quantity & Availability  
10734557
View Documents
Cytiva
28-4038-41
6500 to 75,000
670.00 €
1 set
In Stock
to see stock.
Edge
Add to basket
 
10196234
View Documents
Cytiva
28-4038-42
43,000 to 669,000
670.00 €
1 set
In Stock
to see stock.
Edge
Add to basket
 
Description

Description

  • Optimized range of proteins suited to high-resolution media and prepacked columns
  • Each kit contains five vials of lyophilized proteins
  • Blue Dextran 2000 included to determine the void fraction of the column

Specifications

Specifications

Each
Calibration Kit
2° to 8°C
High-resolution media and prepacked column
Calibration of high-resolution gel filtration media and column such as Superdex™, Superose and Sephacryl
Videos
Safety and Handling

Safety and Handling

Signal Word
  • Danger

Hazard Category
  • Respiratory sensitiser Category 1
  • Skin sensitiser Category 1

Supplemental information
  • MIXTURE LIST-Contains : Trypsin inhibitor, pancreatic basic

SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.