missing translation for 'onlineSavingsMsg'
Learn More

Hamilton™ Gel-Loading Removable Needle Replacements

Designed for multiplex DNA sequencing methodologies.

Brand:  Hamilton™ 8651-01

Product Code. 10768764

  • 278.00 € / Pack of 6

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • Rapid sample transfer
  • Fixed, 25mm long, blunt needles spaced 9mm apart
  • Adjustable volume stop allows faster, more accurate reloading and excellent reproducibility
Specifications

Specifications

Small RN Hub
25.4 mm
6/Pk.
0.2 mm
Gel Loading Removable Needle
3
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers